1-(Ethylthio)-4-(trimethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

168032-5G 168032-25G 168032-1G 168032-500MG
77627-894EA 1599.59 USD
77627-894 77657-648 77640-876 77644-852
1-(Ethylthio)-4-(trimethylsilyl)benzene ≥95%
1-(Ethylthio)-4-(trimethylsilyl)benzene

Precaución: For Research Use Only

Formula: C₁₁H₁₈SSi
Storage Temperature: Ambient
MDL Number: MFCD31923590
CAS Number: 2565807-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR