1-(Ethylthio)-4-phenoxybenzene ≥95%

Supplier: AOBChem USA

Warning

168029-5G 168029-25G 168029-1G 168029-500MG
77631-768EA 1186 USD
77631-768 77657-646 77640-874 77644-850
1-(Ethylthio)-4-phenoxybenzene ≥95%
1-(Ethylthio)-4-phenoxybenzene

Precaución: For Research Use Only

Formula: C₁₄H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD11617693
CAS Number: 19928-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR