1,2-Dichloro-4-(ethylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

168020-5G 168020-25G 168020-1G 168020-500MG
77634-148EA 565 USD
77634-148 77653-808 77640-872 77648-896
1,2-Dichloro-4-(ethylthio)benzene ≥95%
1,2-Dichloro-4-(ethylthio)benzene

Precaución: For Research Use Only

Formula: 207.12 g/mol
MW: C₈H₈Cl₂S
Storage Temperature: Ambient
MDL Number: MFCD11617681
CAS Number: 101079-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR