1-(Ethylthio)-4-methoxy-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

168009-5G 168009-1G 168009-25G 168009-500MG
77631-766EA 1186 USD
77631-766 77636-872 77657-640 77644-848
1-(Ethylthio)-4-methoxy-2-methylbenzene ≥95%
1-(Ethylthio)-4-methoxy-2-methylbenzene

Precaución: For Research Use Only

Formula: 182.28 g/mol
MW: C₁₀H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD11617700
CAS Number: 860734-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR