4-Chloro-2-(ethylthio)-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

167978-1G 167978-500MG 167978-5G
77636-870EA 630 USD
77636-870 77652-406 77653-806
4-Chloro-2-(ethylthio)-1-methylbenzene ≥97%
4-Chloro-2-(ethylthio)-1-methylbenzene

Precaución: For Research Use Only

Formula: 186.7 g/mol
MW: C₉H₁₁ClS
Storage Temperature: Ambient
MDL Number: MFCD11617721
CAS Number: 875236-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR