1-(Ethylthio)-2-methoxynaphthalene ≥97%

Supplier: AOBChem USA

Warning

167970-5G 167970-1G 167970-25G 167970-500MG
77627-888EA 1599.59 USD
77627-888 77636-868 77653-804 77648-892
1-(Ethylthio)-2-methoxynaphthalene ≥97%
1-(Ethylthio)-2-methoxynaphthalene

Precaución: For Research Use Only

Formula: 218.32 g/mol
MW: C₁₃H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD34852404
CAS Number: 100797-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR