4-(Ethylthio)-1,2-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

167969-5G 167969-1G 167969-500MG
77653-802EA 2250 USD
77653-802 77640-870 77648-890
4-(Ethylthio)-1,2-dimethylbenzene ≥95%
4-(Ethylthio)-1,2-dimethylbenzene

Precaución: For Research Use Only

Formula: 166.28 g/mol
MW: C₁₀H₁₄S
Storage Temperature: Ambient
MDL Number: MFCD11617704
CAS Number: 105336-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR