1-(3-Cyclopentyl-4-fluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

167669-5G 167669-1G 167669-500MG
77653-798EA 1811 USD
77653-798 77640-866 77644-846
1-(3-Cyclopentyl-4-fluorophenyl)ethanone ≥95%
1-(3-Cyclopentyl-4-fluorophenyl)ethanone

Precaución: For Research Use Only

Formula: C₁₃H₁₅FO
Storage Temperature: Ambient
CAS Number: 1507258-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR