2-Benzyloxy-3-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16734-5G 16734-1G 16734-25G 16734-500MG
77634-140EA 285 USD
77634-140 77636-866 77657-636 77644-844
2-Benzyloxy-3-fluorobenzaldehyde ≥97%
2-Benzyloxy-3-fluorobenzaldehyde

Precaución: For Research Use Only

Formula: C₁₄H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD11520675
CAS Number: 148872-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR