5-Chloro-α-ethyl-2-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

167066-5G 167066-25G 167066-1G 167066-500MG
77631-762EA 1180 USD
77631-762 77657-634 77640-862 77644-840
5-Chloro-α-ethyl-2-methoxybenzenemethanol ≥95%
5-Chloro-α-ethyl-2-methoxybenzenemethanol

Precaución: For Research Use Only

Formula: 200.66 g/mol
MW: C₁₀H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD11521239
CAS Number: 1215924-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR