2,5-Dichloro-α-ethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

167063-5G 167063-1G 167063-500MG
77627-886EA 1484.23 USD
77627-886 77640-860 77648-882
2,5-Dichloro-α-ethylbenzenemethanol ≥95%
2,5-Dichloro-α-ethylbenzenemethanol

Precaución: For Research Use Only

Formula: 205.08 g/mol
MW: C₉H₁₀Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD12787836
CAS Number: 1249094-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR