3-Chloro-α-ethyl-2-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

167052-5G 167052-25G 167052-1G 167052-500MG
77631-760EA 1180 USD
77631-760 77657-632 77640-858 77644-838
3-Chloro-α-ethyl-2-methylbenzenemethanol ≥95%
3-Chloro-α-ethyl-2-methylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD11521252
CAS Number: 1314904-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR