α-Ethyl-4-fluoro-2-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

167048-5G 167048-25G 167048-1G 167048-500MG
77631-758EA 1180 USD
77631-758 77657-630 77644-834 77644-836
α-Ethyl-4-fluoro-2-methoxybenzenemethanol ≥95%
α-Ethyl-4-fluoro-2-methoxybenzenemethanol

Precaución: For Research Use Only

Formula: 184.21 g/mol
MW: C₁₀H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD11521245
CAS Number: 1314970-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR