α-Ethyl-2-fluoro-3-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

167022-5G 167022-25G 167022-1G 167022-500MG
77627-884EA 1484.23 USD
77627-884 77635-472 77636-862 77648-880
α-Ethyl-2-fluoro-3-methylbenzenemethanol ≥95%
α-Ethyl-2-fluoro-3-methylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD34147575
CAS Number: 1595018-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR