α-Ethyl-2-fluoro-4-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

167020-5G 167020-1G 167020-500MG
77627-882EA 1484.23 USD
77627-882 77640-856 77644-832
α-Ethyl-2-fluoro-4-methylbenzenemethanol ≥95%
α-Ethyl-2-fluoro-4-methylbenzenemethanol

Precaución: For Research Use Only

Formula: 168.21 g/mol
MW: C₁₀H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD30727610
CAS Number: 1596763-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR