α-Ethyl-3-methoxy-4-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

167018-5G 167018-25G 167018-1G 167018-500MG
77627-880EA 1484.23 USD
77627-880 77635-470 77640-854 77648-878
α-Ethyl-3-methoxy-4-methylbenzenemethanol ≥95%
α-Ethyl-3-methoxy-4-methylbenzenemethanol

Precaución: For Research Use Only

Formula: 180.24 g/mol
MW: C₁₁H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD34852124
CAS Number: 1597951-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR