4-Chloro-α-ethyl-2-fluoro-3-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

167011-5G 167011-25G 167011-500MG 167011-1G
77627-878EA 1599.59 USD
77627-878 77635-468 77652-404 77640-852
4-Chloro-α-ethyl-2-fluoro-3-methylbenzenemethanol ≥95%
4-Chloro-α-ethyl-2-fluoro-3-methylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₂ClFO
Storage Temperature: Ambient
MDL Number: MFCD34852120
CAS Number: 1877136-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR