3-Chloro-α-ethyl-4-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166992-5G 166992-25G 166992-1G 166992-500MG
77627-876EA 1841.64 USD
77627-876 77657-628 77640-850 77644-830
3-Chloro-α-ethyl-4-methoxybenzenemethanol ≥95%
3-Chloro-α-ethyl-4-methoxybenzenemethanol

Precaución: For Research Use Only

Formula: 200.66 g/mol
MW: C₁₀H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD12786779
CAS Number: 213470-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR