2,3-Dichloro-α-ethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166990-5G 166990-25G 166990-1G 166990-500MG
77631-756EA 1368 USD
77631-756 77657-626 77640-848 77648-876
2,3-Dichloro-α-ethylbenzenemethanol ≥95%
2,3-Dichloro-α-ethylbenzenemethanol

Precaución: For Research Use Only

Formula: C₉H₁₀Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD17222661
CAS Number: 835925-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR