3-Chloro-α-ethyl-4-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166977-5G 166977-1G 166977-25G 166977-500MG
77631-754EA 1223 USD
77631-754 77636-858 77653-792 77644-828
3-Chloro-α-ethyl-4-methylbenzenemethanol ≥95%
3-Chloro-α-ethyl-4-methylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD11521241
CAS Number: 1270583-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR