α,2-Diethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166969-5G 166969-25G 166969-1G 166969-500MG
77631-752EA 1101 USD
77631-752 77657-624 77640-846 77648-874
α,2-Diethylbenzenemethanol ≥95%
α,2-Diethylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₁H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD11521221
CAS Number: 67541-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR