α-Ethyl-4-phenoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166955-5G 166955-25G 166955-1G 166955-500MG
77627-872EA 1484.23 USD
77627-872 77635-466 77640-844 77648-872
α-Ethyl-4-phenoxybenzenemethanol ≥95%
α-Ethyl-4-phenoxybenzenemethanol

Precaución: For Research Use Only

Formula: 228.29 g/mol
MW: C₁₅H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD16357188
CAS Number: 4974-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR