α-Ethyl-4-(trimethylsilyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166954-1G 166954-5G 166954-500MG
77636-856EA 531 USD
77636-856 77653-790 77648-870
α-Ethyl-4-(trimethylsilyl)benzenemethanol ≥95%
α-Ethyl-4-(trimethylsilyl)benzenemethanol

Precaución: For Research Use Only

Formula: 208.37 g/mol
MW: C₁₂H₂₀OSi
Storage Temperature: Ambient
MDL Number: MFCD34852111
CAS Number: 18052-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR