α-Ethyl-4-methoxy-2-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166952-5G 166952-1G 166952-500MG
77627-870EA 1707.74 USD
77627-870 77636-854 77648-868
α-Ethyl-4-methoxy-2-methylbenzenemethanol ≥95%
α-Ethyl-4-methoxy-2-methylbenzenemethanol

Precaución: For Research Use Only

Formula: 180.24 g/mol
MW: C₁₁H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD11521218
CAS Number: 53773-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR