α-Ethyl-2,3-difluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166950-5G 166950-1G 166950-500MG
77627-868EA 1688.17 USD
77627-868 77640-842 77648-866
α-Ethyl-2,3-difluorobenzenemethanol ≥95%
α-Ethyl-2,3-difluorobenzenemethanol

Precaución: For Research Use Only

Formula: 172.17 g/mol
MW: C₉H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD19602004
CAS Number: 574755-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR