2,4-Dichloro-α-ethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166943-5G 166943-1G 166943-25G 166943-500MG
77631-750EA 1327 USD
77631-750 77636-852 77657-622 77644-826
2,4-Dichloro-α-ethylbenzenemethanol ≥95%
2,4-Dichloro-α-ethylbenzenemethanol

Precaución: For Research Use Only

Formula: 205.08 g/mol
MW: C₉H₁₀Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD08442416
CAS Number: 53066-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR