3,4-Dichloro-α-ethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166942-5G 166942-25G 166942-1G 166942-500MG
77631-748EA 1180 USD
77631-748 77653-786 77640-838 77644-824
3,4-Dichloro-α-ethylbenzenemethanol ≥95%
3,4-Dichloro-α-ethylbenzenemethanol

Precaución: For Research Use Only

Formula: 205.08 g/mol
MW: C₉H₁₀Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD10645145
CAS Number: 53984-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR