4-(1,1-Dimethylethyl)-α-ethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166929-5G 166929-500MG 166929-25G 166929-1G
77634-136EA 528 USD
77634-136 77652-400 77653-784 77640-836
4-(1,1-Dimethylethyl)-α-ethylbenzenemethanol ≥95%
4-(1,1-Dimethylethyl)-α-ethylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₃H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD01070800
CAS Number: 127896-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR