1-[3-(Trifluoromethyl)phenyl]propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(3-(Trifluoromethyl)phenyl)propan-1-ol

166928-5G 166928-25G 166928-1G 166928-500MG
77634-134EA 329 USD
77634-134 77657-620 77640-834 77648-862
1-[3-(Trifluoromethyl)phenyl]propan-1-ol ≥95%
1-[3-(Trifluoromethyl)phenyl]propan-1-ol

Precaución: For Research Use Only

Formula: C₁₀H₁₁F₃O
MW: 204.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046668
CAS Number: 618-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR