1-(3,4-Dimethoxyphenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

166916-5G 166916-25G 166916-1G 166916-500MG
77631-744EA 670 USD
77631-744 77653-780 77640-832 77644-820
1-(3,4-Dimethoxyphenyl)propan-1-ol ≥95%
1-(3,4-Dimethoxyphenyl)propan-1-ol

Precaución: For Research Use Only

Formula: C₁₁H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD01117649
CAS Number: 10548-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR