α-Ethyl-α-methyl-2-(methylthio)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

166866-5G 166866-1G 166866-500MG
77653-776EA 2250 USD
77653-776 77640-830 77644-818
α-Ethyl-α-methyl-2-(methylthio)benzenemethanol ≥97%
α-Ethyl-α-methyl-2-(methylthio)benzenemethanol

Precaución: For Research Use Only

Formula: 196.31 g/mol
MW: C₁₁H₁₆OS
Storage Temperature: Ambient
MDL Number: MFCD20528406
CAS Number: 1379300-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR