(4-Cyclopentylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

166593-5G 166593-1G 166593-500MG 166593-25G
77631-742EA 1441 USD
77631-742 77636-846 77652-398 77653-774
(4-Cyclopentylphenyl)(methyl)sulfane ≥95%
(4-Cyclopentylphenyl)(methyl)sulfane

Precaución: For Research Use Only

Formula: 192.3204 g/mol
MW: C₁₂H₁₆S
Storage Temperature: Ambient
MDL Number: MFCD34851979
CAS Number: 2921719-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR