3-Fluoro-2-isopropoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

16646-5G 16646-1G 16646-25G 16646-500MG
77634-130EA 384 USD
77634-130 77636-844 77657-618 77644-816
3-Fluoro-2-isopropoxybenzaldehyde ≥95%
3-Fluoro-2-isopropoxybenzaldehyde

Precaución: For Research Use Only

Formula: 182.192 g/mol
MW: C₁₀H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD26056440
CAS Number: 1037092-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR