1-Cyclopentyl-3,5-difluorobenzene ≥95%

Supplier: AOBChem USA

Warning

166452-1G 166452-5G 166452-500MG
77636-842EA 626 USD
77636-842 77657-616 77652-396
1-Cyclopentyl-3,5-difluorobenzene ≥95%
1-Cyclopentyl-3,5-difluorobenzene

Precaución: For Research Use Only

Formula: C₁₁H₁₂F₂
Storage Temperature: Ambient
MDL Number: MFCD26678754
CAS Number: 1516624-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR