3-[2-Methyl-3-(trifluoromethyl)phenyl]-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166374-1G 166374-5G 166374-500MG
77636-840EA 580 USD
77636-840 77657-612 77644-814
3-[2-Methyl-3-(trifluoromethyl)phenyl]-3-oxetanol ≥95%
3-[2-Methyl-3-(trifluoromethyl)phenyl]-3-oxetanol

Precaución: For Research Use Only

Formula: 232.2 g/mol
MW: C₁₁H₁₁F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD24396205
CAS Number: 1432620-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR