3-[3-(Trifluoromethyl)phenyl]-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166365-5G 166365-25G 166365-1G 166365-500MG
77627-866EA 1484.23 USD
77627-866 77653-768 77640-826 77644-810
3-[3-(Trifluoromethyl)phenyl]-3-oxetanol ≥95%
3-[3-(Trifluoromethyl)phenyl]-3-oxetanol

Precaución: For Research Use Only

Formula: 218.17 g/mol
MW: C₁₀H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD28604140
CAS Number: 1528651-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR