3-(3,4-Dimethylphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166358-5G 166358-1G 166358-500MG
77627-864EA 1484.23 USD
77627-864 77636-836 77644-808
3-(3,4-Dimethylphenyl)-3-oxetanol ≥95%
3-(3,4-Dimethylphenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: 178.23 g/mol
MW: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34851903
CAS Number: 1889495-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR