3-(2,5-Difluorophenyl)-3-oxetanol ≥97%

Supplier: AOBChem USA

Warning

166351-5G 166351-1G 166351-500MG
77627-862EA 1484.23 USD
77627-862 77636-834 77648-858
3-(2,5-Difluorophenyl)-3-oxetanol ≥97%
3-(2,5-Difluorophenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: C₉H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34851896
CAS Number: 1894507-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR