3-(2,5-Dimethylphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166345-5G 166345-1G 166345-500MG
77627-860EA 1615.04 USD
77627-860 77640-822 77648-856
3-(2,5-Dimethylphenyl)-3-oxetanol ≥95%
3-(2,5-Dimethylphenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: 178.23 g/mol
MW: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34851891
CAS Number: 1903401-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR