3-(3-Ethylphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166334-5G 166334-1G 166334-500MG
77627-858EA 1609.89 USD
77627-858 77636-832 77652-394
3-(3-Ethylphenyl)-3-oxetanol ≥95%
3-(3-Ethylphenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: 178.23 g/mol
MW: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34851880
CAS Number: 2103941-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR