3-(3-Chloro-4-methylphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166329-5G 166329-1G 166329-500MG
77657-608EA 2422 USD
77657-608 77640-820 77644-804
3-(3-Chloro-4-methylphenyl)-3-oxetanol ≥95%
3-(3-Chloro-4-methylphenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: C₁₀H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34851875
CAS Number: 2104646-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR