3-(3,5-Dimethylphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166326-1G 166326-5G 166326-500MG
77636-830EA 518 USD
77636-830 77657-606 77644-802
3-(3,5-Dimethylphenyl)-3-oxetanol ≥95%
3-(3,5-Dimethylphenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: 178.23 g/mol
MW: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34851872
CAS Number: 2104720-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR