3-[3-(1-Methylethyl)phenyl]-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166324-5G 166324-1G 166324-500MG
77627-856EA 1484.23 USD
77627-856 77636-828 77648-854
3-[3-(1-Methylethyl)phenyl]-3-oxetanol ≥95%
3-[3-(1-Methylethyl)phenyl]-3-oxetanol

Precaución: For Research Use Only

Formula: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD34851870
CAS Number: 2105007-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR