3-(5-Fluoro-2-methylphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166319-5G 166319-1G 166319-500MG
77627-854EA 1616.07 USD
77627-854 77636-826 77652-392
3-(5-Fluoro-2-methylphenyl)-3-oxetanol ≥95%
3-(5-Fluoro-2-methylphenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: 182.19 g/mol
MW: C₁₀H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD34851865
CAS Number: 2106973-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR