3-(3-Fluoro-5-methoxyphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166315-5G 166315-1G 166315-25G 166315-500MG
77627-852EA 1484.23 USD
77627-852 77636-824 77657-604 77644-800
3-(3-Fluoro-5-methoxyphenyl)-3-oxetanol ≥95%
3-(3-Fluoro-5-methoxyphenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: 198.19 g/mol
MW: C₁₀H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD34851861
CAS Number: 2107711-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR