3-(2-Ethylphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166308-1G 166308-5G 166308-500MG
77636-822EA 518 USD
77636-822 77653-764 77644-798
3-(2-Ethylphenyl)-3-oxetanol ≥95%
3-(2-Ethylphenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34851854
CAS Number: 2110286-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR