3-(2,3-Dichlorophenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166306-5G 166306-1G 166306-500MG
77627-850EA 1707.74 USD
77627-850 77640-816 77644-794
3-(2,3-Dichlorophenyl)-3-oxetanol ≥95%
3-(2,3-Dichlorophenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: C₉H₈Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34851852
CAS Number: 2110944-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR