3-(4-Chloro-2-methylphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166302-5G 166302-1G 166302-500MG
77627-848EA 1484.23 USD
77627-848 77640-814 77648-852
3-(4-Chloro-2-methylphenyl)-3-oxetanol ≥95%
3-(4-Chloro-2-methylphenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: 198.65 g/mol
MW: C₁₀H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34851848
CAS Number: 2149783-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR