3-(2-Chloro-4-methylphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166299-500MG 166299-5G 166299-1G
77652-390EA 444 USD
77652-390 77653-760 77640-812
3-(2-Chloro-4-methylphenyl)-3-oxetanol ≥95%
3-(2-Chloro-4-methylphenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: C₁₀H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD30699856
CAS Number: 2150938-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR