3-[2-(Trifluoromethyl)phenyl]-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166295-5G 166295-1G 166295-500MG
77653-758EA 1811 USD
77653-758 77640-810 77648-850
3-[2-(Trifluoromethyl)phenyl]-3-oxetanol ≥95%
3-[2-(Trifluoromethyl)phenyl]-3-oxetanol

Precaución: For Research Use Only

Formula: 218.17 g/mol
MW: C₁₀H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34851842
CAS Number: 2161156-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR